Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114479.5 | NP_190196.1 | 4 - 5 | 669 | 106 | |
GenBank mRNA | AF020715.1 | - | 4 - 5 | 605 | 106 | |
AY056233.1 | - | 4 - 5 | 610 | 106 | ||
AY117252.1 | - | 4 - 5 | 600 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |