Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339245.1 | NP_001326106.1 | 9 - 10 | 2070 | 153 | |
NM_114500.5 | NP_190217.2 | 10 - 11 | 2142 | 153 | ||
GenBank mRNA | FJ708732.1 | - | 10 - 11 | 1957 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |