Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114573.3 | NP_566889.1 | 1 - 2 | 894 | 64 | |
GenBank mRNA | AY099737.1 | - | 1 - 2 | 835 | 64 | |
BT000368.1 | - | 1 - 2 | 821 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |