Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114641.4 | NP_190357.2 | 13 - 14 | 2625 | 138 | |
GenBank mRNA | BT004197.1 | - | 13 - 14 | 2438 | 138 | |
BT005456.1 | - | 13 - 14 | 2370 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |