Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339337.1 | NP_001327142.1 | 15 - 16 | 2665 | 86 | |
NM_001339338.1 | NP_001327141.1 | 16 - 17 | 2574 | 86 | ||
NM_114647.4 | NP_190363.3 | 17 - 18 | 2483 | 86 | ||
GenBank mRNA | AY099665.1 | - | 17 - 18 | 2482 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |