Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339337.1 | NP_001327142.1 | 7 - 8 | 1310 | 96 | |
NM_001339338.1 | NP_001327141.1 | 8 - 9 | 1219 | 96 | ||
NM_114647.4 | NP_190363.3 | 8 - 9 | 1219 | 96 | ||
GenBank mRNA | AY099665.1 | - | 8 - 9 | 1218 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |