Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114669.4 | NP_190383.1 | 4 - 5 | 841 | 120 | |
GenBank mRNA | AB030820.1 | - | 4 - 5 | 597 | 120 | |
AF327426.1 | - | 4 - 5 | 591 | 120 | ||
AF349447.1 | - | 4 - 5 | 597 | 120 | ||
AF349522.1 | - | 4 - 5 | 515 | 120 | ||
AF372911.1 | - | 4 - 5 | 591 | 120 | ||
AY090443.1 | - | 4 - 5 | 595 | 120 | ||
BT000752.1 | - | 4 - 5 | 540 | 120 | ||
BT006371.1 | - | 4 - 5 | 515 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |