Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114670.2 | NP_190384.1 | 2 - 3 | 807 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |