Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP94B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114710.3 | NP_190421.1 | 1 | 1455 | 65 | |
GenBank mRNA | AK226339.1 | - | 1 | 1352 | 65 | |
BT015332.1 | - | 1 | 1347 | 65 | ||
BT015841.1 | - | 1 | 1305 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |