Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114732.5 | NP_190442.1 | 3 - 4 | 1118 | 75 | |
GenBank mRNA | AK229328.1 | - | 3 - 4 | 1066 | 75 | |
BT025324.1 | - | 3 - 4 | 992 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |