Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT3;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114744.4 | NP_190454.1 | 1 - 2 | 438 | 101 | |
GenBank mRNA | BT033111.1 | - | 1 - 2 | 289 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |