Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP93-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203106.2 | NP_001190035.1 | 8 - 9 | 1738 | 76 | |
NM_001339385.1 | NP_001325472.1 | 8 - 9 | 1880 | 76 | ||
NM_001339386.1 | NP_001325473.1 | 9 - 10 | 1830 | 76 | ||
NM_114746.5 | NP_566912.2 | 8 - 9 | 1780 | 76 | ||
GenBank mRNA | AB022324.1 | - | 8 - 9 | 1658 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |