Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114784.3 | NP_566916.1 | 3 - 4 | 705 | 67 | |
GenBank mRNA | AY049288.1 | - | 3 - 4 | 653 | 67 | |
AY090286.1 | - | 3 - 4 | 510 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |