Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CID12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036537.1 | NP_001031614.1 | 4 - 5 | 756 | 70 | |
NM_117129.3 | NP_192799.1 | 4 - 5 | 951 | 70 | ||
GenBank mRNA | AY039599.1 | - | 4 - 5 | 695 | 70 | |
AY078011.1 | - | 4 - 5 | 597 | 70 | ||
AY086286.1 | - | 4 - 5 | 673 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |