Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340684.1 | NP_001328829.1 | 4 - 5 | 1147 | 82 | |
NM_117145.3 | NP_192815.1 | 5 - 6 | 1108 | 82 | ||
GenBank mRNA | AY045668.1 | - | 5 - 6 | 1032 | 82 | |
BT004519.1 | - | 5 - 6 | 949 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |