Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTSSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117176.3 | NP_192844.2 | 5 - 6 | 761 | 146 | |
GenBank mRNA | AJ582222.1 | - | 4 - 5 | 533 | 146 | |
BT004166.1 | - | 5 - 6 | 698 | 146 | ||
BT005015.1 | - | 4 - 5 | 533 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |