Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340739.1 | NP_001319907.1 | 5 - 6 | 746 | 101 | |
NM_117258.5 | NP_192925.1 | 5 - 6 | 553 | 101 | ||
GenBank mRNA | AK230464.1 | - | 5 - 6 | 554 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |