Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341318.1 | NP_001329724.1 | 5 - 6 | 1588 | 114 | |
NM_118020.5 | NP_193637.2 | 4 - 5 | 1767 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |