Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341876.1 | NP_001329759.1 | 3 - 4 | 449 | 63 | |
NM_001341877.1 | NP_001329760.1 | 2 - 3 | 607 | 63 | ||
NM_118901.5 | NP_194495.3 | 3 - 4 | 438 | 63 | ||
GenBank mRNA | AF069497.1 | - | 3 - 4 | 333 | 63 | |
AY074298.1 | - | 3 - 4 | 312 | 63 | ||
AY096569.1 | - | 2 - 3 | 132 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |