Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036655.1 | NP_001031732.1 | 2 - 3 | 181 | 89 | |
NM_118908.5 | NP_194499.2 | 4 - 5 | 369 | 89 | ||
GenBank mRNA | AY094409.1 | - | 4 - 5 | 282 | 89 | |
AY122891.1 | - | 2 - 3 | 155 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |