Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DD46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102051.3 | NP_564154.1 | 6 - 7 | 687 | 136 | |
GenBank mRNA | AK226416.1 | - | 6 - 7 | 688 | 136 | |
KJ138995.1 | - | 6 - 7 | 593 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |