Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332533.1 | NP_001323098.1 | 1 - 2 | 1388 | 92 | |
NM_102052.3 | NP_173621.1 | 1 - 2 | 1365 | 92 | ||
GenBank mRNA | AY125514.1 | - | 1 - 2 | 1348 | 92 | |
BT004532.1 | - | 1 - 2 | 1070 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |