Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT4;5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343659.1 | NP_001330368.1 | 12 - 13 | 1517 | 66 | |
NM_122045.4 | NP_197538.2 | 12 - 13 | 1405 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |