Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161252.2 | NP_001154724.1 | 5 - 6 | 655 | 69 | |
NM_001343670.1 | NP_001318611.1 | 5 - 6 | 634 | 69 | ||
NM_001343671.1 | NP_001318612.1 | 6 - 7 | 1087 | 69 | ||
GenBank mRNA | HQ427882.1 | - | 5 - 6 | 637 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |