Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAZ12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122098.5 | NP_197590.1 | 4 - 5 | 643 | 100 | |
GenBank mRNA | AF360184.1 | - | 4 - 5 | 558 | 100 | |
AY051013.1 | - | 4 - 5 | 464 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |