Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122101.2 | NP_568405.1 | 14 - 15 | 2155 | 102 | |
GenBank mRNA | L23985.1 | - | 14 - 15 | 2115 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |