Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122140.4 | NP_568417.1 | 3 - 4 | 1060 | 92 | |
GenBank mRNA | AB379589.1 | - | 3 - 4 | 516 | 92 | |
AY059915.1 | - | 3 - 4 | 708 | 92 | ||
AY128808.1 | - | 3 - 4 | 516 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |