Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036849.2 | NP_001031926.1 | 4 - 5 | 472 | 60 | |
NM_122214.4 | NP_197699.2 | 4 - 5 | 559 | 60 | ||
GenBank mRNA | AK118129.1 | - | 4 - 5 | 467 | 60 | |
AY518689.1 | - | 4 - 5 | 428 | 60 | ||
BT008588.1 | - | 4 - 5 | 428 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |