Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122214.4 | NP_197699.2 | 8 - 9 | 901 | 92 | |
GenBank mRNA | AK118129.1 | - | 8 - 9 | 809 | 92 | |
BT008588.1 | - | 8 - 9 | 770 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |