Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122360.2 | NP_197840.1 | 1 - 2 | 1135 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |