Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCA2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343923.1 | NP_001318646.1 | 8 - 9 | 1156 | 68 | |
NM_001343924.1 | NP_001332474.1 | 8 - 9 | 1156 | 68 | ||
NM_001343925.1 | NP_001332473.1 | 7 - 8 | 958 | 68 | ||
NM_122447.3 | NP_197920.2 | 7 - 8 | 940 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |