Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLC-D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343969.1 | NP_001330038.1 | 20 - 21 | 2442 | 70 | |
NM_122525.3 | NP_197996.1 | 20 - 21 | 2457 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |