Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK 11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122557.3 | NP_568486.1 | 2 - 3 | 347 | 113 | |
GenBank mRNA | AF428327.1 | - | 2 - 3 | 347 | 113 | |
AY046024.1 | - | 2 - 3 | 347 | 113 | ||
AY142595.1 | - | 1 - 2 | 104 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |