Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102057.4 | NP_564156.1 | 2 - 3 | 717 | 84 | |
GenBank mRNA | BT026034.1 | - | 1 - 2 | 199 | 84 | |
L10209.1 | - | 2 - 3 | 280 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |