Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT85A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102087.2 | NP_564170.1 | 1 | 2207 | 120 | |
NM_202156.2 | NP_973885.1 | 2 | 1883 | 120 | ||
GenBank mRNA | AY039897.1 | - | 1 | 1134 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |