Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102118.4 | NP_173685.1 | 3 - 4 | 1554 | 121 | |
GenBank mRNA | AY048256.1 | - | 3 - 4 | 1441 | 121 | |
AY050986.1 | - | 3 - 4 | 1449 | 121 | ||
AY088566.1 | - | 3 - 4 | 1449 | 121 | ||
AY113946.1 | - | 3 - 4 | 1368 | 121 | ||
BT000684.1 | - | 3 - 4 | 1515 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |