Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102141.3 | NP_173707.2 | 6 - 7 | 838 | 92 | |
GenBank mRNA | BT020417.1 | - | 6 - 7 | 793 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |