Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332622.1 | NP_001321290.1 | 6 - 7 | 904 | 108 | |
NM_102228.4 | NP_564204.1 | 6 - 7 | 904 | 108 | ||
GenBank mRNA | AY035139.1 | - | 6 - 7 | 896 | 108 | |
AY113912.1 | - | 6 - 7 | 822 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |