Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102258.6 | NP_173822.2 | 1 - 2 | 452 | 120 | |
GenBank mRNA | AY074564.1 | - | 1 - 2 | 379 | 120 | |
AY226825.1 | - | 1 - 2 | 384 | 120 | ||
BT025997.1 | - | 1 - 2 | 136 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |