Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198151.2 | NP_001185080.1 | 4 - 5 | 646 | 82 | |
NM_001332641.1 | NP_001323398.1 | 4 - 5 | 602 | 82 | ||
NM_102265.3 | NP_173829.3 | 4 - 5 | 571 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |