Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102274.3 | NP_173838.1 | 8 - 9 | 1505 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |