Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102352.1 | NP_173912.1 | 1 | 979 | 73 | |
GenBank mRNA | AB062612.1 | - | 1 | 979 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |