Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102360.4 | NP_173920.1 | 2 - 3 | 573 | 115 | |
GenBank mRNA | AY120757.1 | - | 2 - 3 | 515 | 115 | |
BT000108.1 | - | 1 - 2 | 398 | 115 | ||
U21557.1 | - | 2 - 3 | 476 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |