Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM20-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102502.3 | NP_174059.2 | 1 - 2 | 241 | 90 | |
GenBank mRNA | AK228213.1 | - | 1 - 2 | 192 | 90 | |
BT024602.1 | - | 1 - 2 | 103 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |