Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUB45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102556.5 | NP_174112.1 | 3 - 4 | 673 | 105 | |
GenBank mRNA | AY094469.1 | - | 3 - 4 | 585 | 105 | |
BT002721.1 | - | 2 - 3 | 241 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |