Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123901.2 | NP_001117373.2 | 11 - 12 | 1180 | 142 | |
NM_001332819.1 | NP_001321583.1 | 9 - 10 | 1230 | 142 | ||
NM_102624.4 | NP_174178.2 | 10 - 11 | 1147 | 142 | ||
GenBank mRNA | AY099624.1 | - | 10 - 11 | 1058 | 142 | |
BT003404.1 | - | 7 - 8 | 590 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |