Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAVE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123903.2 | NP_001117375.1 | 2 - 3 | 618 | 145 | |
NM_001123904.2 | NP_001117376.1 | 2 - 3 | 591 | 145 | ||
NM_102658.3 | NP_174212.2 | 2 - 3 | 618 | 145 | ||
GenBank mRNA | AY743925.1 | - | 2 - 3 | 305 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |