Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106062.4 | NP_177543.1 | 5 - 6 | 1547 | 73 | |
GenBank mRNA | AY034978.1 | - | 5 - 6 | 1347 | 73 | |
AY062968.1 | - | 5 - 6 | 1316 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |