Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344021.1 | NP_001331944.1 | 8 - 9 | 1067 | 98 | |
NM_001344022.1 | NP_001331946.1 | 8 - 9 | 1067 | 98 | ||
NM_001344023.1 | NP_001331945.1 | 8 - 9 | 898 | 98 | ||
NM_122645.5 | NP_198115.2 | 8 - 9 | 924 | 98 | ||
GenBank mRNA | AY093156.1 | - | 8 - 9 | 898 | 98 | |
BT008399.1 | - | 8 - 9 | 820 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |