Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036891.2 | NP_001031968.1 | 2 - 3 | 540 | 74 | |
NM_001203493.1 | NP_001190422.1 | 2 - 3 | 592 | 74 | ||
NM_122927.4 | NP_198386.1 | 2 - 3 | 594 | 74 | ||
GenBank mRNA | AY085968.1 | - | 2 - 3 | 537 | 74 | |
AY142630.1 | - | 2 - 3 | 398 | 74 | ||
U90879.1 | - | 2 - 3 | 516 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |